Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals
Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459640050
Price (USD)/ Each
324.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
382.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
324.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
382.00 
Save 57.35 (15%)
Add to cart
Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals
Catalog number459640050
Price (USD)/ Each
324.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
382.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
CAS754-91-6
IUPAC Nameheptadecafluorooctane-1-sulfonamide
Molecular FormulaC8H2F17NO2S
InChI KeyRRRXPPIDPYTNJG-UHFFFAOYSA-N
SMILESNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Solid
GC>=94 % (mix of isomers)
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only