Silver sulfadiazine, 98%, Thermo Scientific Chemicals
Silver sulfadiazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver sulfadiazine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459650250
Price (USD)/ Each
-
Quantity:
25g
Request bulk or custom format
Chemical Identifiers
IUPAC Namesilver(1+) 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
Molecular FormulaC10H9AgN4O2S
InChI KeyUEJSSZHHYBHCEL-UHFFFAOYSA-N
SMILES[Ag+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1
Molecular Weight (g/mol)357.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<0.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only