Thermo Scientific Chemicals

Hexyl nicotinate, 98%, Thermo Scientific Chemicals

Catalog number: 460090250
25 g, Each
Thermo Scientific Chemicals

Hexyl nicotinate, 98%, Thermo Scientific Chemicals

Catalog number: 460090250
25 g, Each
Quantity
Catalog number: 460090250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23597-82-2
IUPAC Name
hexyl pyridine-3-carboxylate
Molecular Formula
C12H17NO2
InChI Key
RVYGVBZGSFLJKH-UHFFFAOYSA-N
SMILES
CCCCCCOC(=O)C1=CC=CN=C1
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4890 to 1.4910 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text