Thermo Scientific Chemicals

Efaproxiral sodium, 98%

Catalog number: 460102500
250 mg, Each
Thermo Scientific Chemicals

Efaproxiral sodium, 98%

Catalog number: 460102500
250 mg, Each
Quantity
Catalog number: 460102500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
170787-99-2
IUPAC Name
sodium 2-(4-{[(3,5-dimethylphenyl)carbamoyl]methyl}phenoxy)-2-methylpropanoate
Molecular Formula
C20H22NNaO4
InChI Key
SWDPIHPGORBMFR-UHFFFAOYSA-M
SMILES
[Na+].CC1=CC(NC(=O)CC2=CC=C(OC(C)(C)C([O-])=O)C=C2)=CC(C)=C1
Loss on drying
=<13 %
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text