Thermo Scientific Chemicals

Succinylcholine chloride, 96%, Thermo Scientific Chemicals

Catalog number: 460110050
5 g, Each
Thermo Scientific Chemicals

Succinylcholine chloride, 96%, Thermo Scientific Chemicals

Catalog number: 460110050
5 g, Each
Quantity
Catalog number: 460110050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
71-27-2
IUPAC Name
trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dichloride
Molecular Formula
C14H30Cl2N2O4
InChI Key
YOEWQQVKRJEPAE-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C
Melting point
160°C to 166°C
Titration Argentometric
>=95 % (on dry substance)
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text