Thermo Scientific Chemicals

Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals

Catalog number: 460390010
1 g, Each
Thermo Scientific Chemicals

Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals

Catalog number: 460390010
1 g, Each
Quantity
Catalog number: 460390010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-39-0
IUPAC Name
methyl hexadecanoate
Molecular Formula
C17H34O2
InChI Key
FLIACVVOZYBSBS-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OC
Infrared spectrum
Conforms
Appearance (Form)
Low melting solid
Appearance (Color)
White
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text