Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 460390050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS112-39-0
IUPAC Namemethyl hexadecanoate
Molecular FormulaC17H34O2
InChI KeyFLIACVVOZYBSBS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Low melting solid
Appearance (Color)White
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only