Chemical Identifiers
CAS552-66-9
IUPAC Name3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Molecular FormulaC21H20O9
InChI KeyKYQZWONCHDNPDP-QNDFHXLGSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
View more