Chemical Identifiers
CAS56-47-3
IUPAC Name2-[(1S,3aS,3bS,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Molecular FormulaC23H32O4
InChI KeyVPGRYOFKCNULNK-ZIUYWEQENA-N
SMILES[H][C@@]12CC[C@H](C(=O)COC(C)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])CCC2=CC(=O)CC[C@]12C
View more