Thermo Scientific Chemicals

N4-Benzoylcytosine, 98%, Thermo Scientific Chemicals

Catalog number: 460540050
5 g, Each
Thermo Scientific Chemicals

N4-Benzoylcytosine, 98%, Thermo Scientific Chemicals

Catalog number: 460540050
5 g, Each
Quantity
Catalog number: 460540050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26661-13-2
IUPAC Name
N-(2-oxo-2,3-dihydropyrimidin-4-yl)benzamide
Molecular Formula
C11H9N3O2
InChI Key
XBDUZBHKKUFFRH-UHFFFAOYSA-N
SMILES
O=C(NC1=CC=NC(=O)N1)C1=CC=CC=C1
Appearance (Form)
Powder or crystalline powder
Elemental Analysis
59.86 to 62.93 % Carbon
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Elemental Analysis
19.04 to 20.01 % Nitrogen

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text