Thermo Scientific Chemicals

2-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 460550250
25 g, Each
Thermo Scientific Chemicals

2-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 460550250
25 g, Each
Quantity
Catalog number: 460550250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22927-13-5
IUPAC Name
2-ethylbenzaldehyde
Molecular Formula
C9H10O
InChI Key
NTWBHJYRDKBGBR-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1C=O
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text