Thermo Scientific Chemicals

3,5-Dibromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 460560250
25 g, Each
Thermo Scientific Chemicals

3,5-Dibromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 460560250
25 g, Each
Quantity
Catalog number: 460560250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
626-40-4
IUPAC Name
3,5-dibromoaniline
Molecular Formula
C6H5Br2N
InChI Key
RVNUUWJGSOHMRR-UHFFFAOYSA-N
SMILES
NC1=CC(Br)=CC(Br)=C1
Appearance (Form)
Solid or crystalline solid
Appearance (Color)
Brown to black
Infrared spectrum
Conforms
Melting point
51°C to 57°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text