Thermo Scientific Chemicals

Budesonide, 97%, Thermo Scientific Chemicals

Catalog number: 460620050
5g, Each
Thermo Scientific Chemicals

Budesonide, 97%, Thermo Scientific Chemicals

Catalog number: 460620050
5g, Each
Quantity
Catalog number: 460620050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51333-22-3
IUPAC Name
(1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Molecular Formula
C25H34O6
InChI Key
VOVIALXJUBGFJZ-VXKMTNQYSA-N
SMILES
CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Infrared spectrum
Conforms
Loss on drying
=<0.5 %
Appearance (Form)
Powder
HPLC
>=96.0 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text