Thermo Scientific Chemicals

2,4-Dichloroquinazoline, 98%, Thermo Scientific Chemicals

Catalog number: 460730050
5 g, Each
Thermo Scientific Chemicals

2,4-Dichloroquinazoline, 98%, Thermo Scientific Chemicals

Catalog number: 460730050
5 g, Each
Quantity
Catalog number: 460730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
607-68-1
IUPAC Name
2,4-dichloroquinazoline
Molecular Formula
C8H4Cl2N2
InChI Key
TUQSVSYUEBNNKQ-UHFFFAOYSA-N
SMILES
ClC1=NC(Cl)=C2C=CC=CC2=N1
Appearance (Color)
White to yellow to light brown
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Powder or crystalline powder
Melting point
116°C to 123°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text