Thermo Scientific Chemicals

Triethylamine, for HPLC, Thermo Scientific Chemicals

Catalog number: 460812500
250 mL, Each
Thermo Scientific Chemicals

Triethylamine, for HPLC, Thermo Scientific Chemicals

Catalog number: 460812500
250 mL, Each
Quantity
Catalog number: 460812500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-44-8
IUPAC Name
triethylamine
Molecular Formula
C6H15N
InChI Key
ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES
CCN(CC)CC
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Water
=<0.05 %
Lead (Pb)
=<0.02 ppm
UV
at 254 nm A: =<0.125 (0.1 M in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text