Thermo Scientific Chemicals

Azaperone, Thermo Scientific Chemicals

Catalog number: 460830250
25 g, Each
Thermo Scientific Chemicals

Azaperone, Thermo Scientific Chemicals

Catalog number: 460830250
25 g, Each
Quantity
Catalog number: 460830250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1649-18-9
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (60°C, 4 h) (vacuum)
Appearance (Form)
Powder
Melting point
92°C to 95°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text