Thermo Scientific Chemicals

Lamotrigine, 98%, Thermo Scientific Chemicals

Catalog number: 460850010
1 g, Each
Thermo Scientific Chemicals

Lamotrigine, 98%, Thermo Scientific Chemicals

Catalog number: 460850010
1 g, Each
Quantity
Catalog number: 460850010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84057-84-1
IUPAC Name
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Molecular Formula
C9H7Cl2N5
InChI Key
PYZRQGJRPPTADH-UHFFFAOYSA-N
SMILES
NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
Loss on drying
=<0.5 % (105°C, 3 h)
HPLC
>=97.5 %
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text