Thermo Scientific Chemicals

Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 460860010
1 g, Each
Thermo Scientific Chemicals

Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 460860010
1 g, Each
Quantity
Catalog number: 460860010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70024-40-7
IUPAC Name
hydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine dihydrate chloride
Molecular Formula
C19H30ClN5O6
InChI Key
NZMOFYDMGFQZLS-UHFFFAOYNA-N
SMILES
[H+].O.O.[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
HPLC
>=97.5 %
Loss on drying
=<9 % (105°C, 3 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text