Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals
Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals
Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals
Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4608600505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460860050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS70024-40-7
IUPAC Namehydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine dihydrate chloride
Molecular FormulaC19H30ClN5O6
InChI KeyNZMOFYDMGFQZLS-UHFFFAOYNA-N
SMILES[H+].O.O.[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Appearance (Form)Powder
HPLC>=97.5 %
Loss on drying=<9 % (105°C, 3 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link