Pantoprazole sodium salt hydrate
Pantoprazole sodium salt hydrate
Pantoprazole sodium salt hydrate
Pantoprazole sodium salt hydrate
Thermo Scientific Chemicals

Pantoprazole sodium salt hydrate

CAS: 718635-09-7 | C16H14F2N3NaO4S | 405.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460870010
Price (USD)
42.65
Special offer
Online exclusive
Ends: 26-Sep-2025
52.60
Save 9.95 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 26-Sep-2025
52.60
Save 9.95 (19%)
Each
Add to cart
Chemical Identifiers
CAS718635-09-7
IUPAC Namesodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazol-1-ide
Molecular FormulaC16H14F2N3NaO4S
InChI KeyYNWDKZIIWCEDEE-UHFFFAOYNA-N
SMILES[Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water4 to 6 %
Titration with HClO498 to 102 %
Solubility(1 % in water) Clear solution
Infrared spectrumConforms
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection