Benzbromarone, 98%, Thermo Scientific Chemicals
Benzbromarone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzbromarone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 460900010
Price (USD)/ Each
47.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
47.20
Add to cart
Benzbromarone, 98%, Thermo Scientific Chemicals
Catalog number460900010
Price (USD)/ Each
47.20
-
Add to cart
Chemical Identifiers
IUPAC Name2,6-dibromo-4-(2-ethyl-1-benzofuran-3-carbonyl)phenol
Molecular FormulaC17H12Br2O3
InChI KeyWHQCHUCQKNIQEC-UHFFFAOYSA-N
SMILESCCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=CC=C2O1
Molecular Weight (g/mol)424.09
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 %
HPLC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only