Thermo Scientific Chemicals

Benzbromarone, 98%, Thermo Scientific Chemicals

Catalog number: 460900050
5 g, Each
Thermo Scientific Chemicals

Benzbromarone, 98%, Thermo Scientific Chemicals

Catalog number: 460900050
5 g, Each
Quantity
Catalog number: 460900050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3562-84-3
IUPAC Name
2,6-dibromo-4-(2-ethyl-1-benzofuran-3-carbonyl)phenol
Molecular Formula
C17H12Br2O3
InChI Key
WHQCHUCQKNIQEC-UHFFFAOYSA-N
SMILES
CCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=CC=C2O1
Loss on drying
=<0.5 %
HPLC
>=97.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text