Thermo Scientific Chemicals

Sodium diatrizoate, 99%, Thermo Scientific Chemicals

Catalog number: 460960250
25 g, Each
Thermo Scientific Chemicals

Sodium diatrizoate, 99%, Thermo Scientific Chemicals

Catalog number: 460960250
25 g, Each
Quantity
Catalog number: 460960250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
737-31-5
IUPAC Name
sodium 3,5-diacetamido-2,4,6-triiodobenzoate
Molecular Formula
C11H8I3N2NaO4
InChI Key
ZEYOIOAKZLALAP-UHFFFAOYSA-M
SMILES
[Na+].CC(=O)NC1=C(I)C(C([O-])=O)=C(I)C(NC(C)=O)=C1I
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=98.5 %
Loss on drying
=<8 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text