Thermo Scientific Chemicals

(S)-(+)-Clopidogrel hydrogensulfate, 98%

Catalog number: 461030010
1 g, Each
Thermo Scientific Chemicals

(S)-(+)-Clopidogrel hydrogensulfate, 98%

Catalog number: 461030010
1 g, Each
Quantity
Catalog number: 461030010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120202-66-6
Specific optical rotation
+54° to +58° (20°C, 589 nm) (1 % in Methanol)
Appearance (Color)
White to beige
Loss on drying
=<0.5 %
HPLC
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text