Thermo Scientific Chemicals

Medroxyprogesterone acetate, 97%, Thermo Scientific Chemicals

Catalog number: 461120050
5g, Each
Thermo Scientific Chemicals

Medroxyprogesterone acetate, 97%, Thermo Scientific Chemicals

Catalog number: 461120050
5g, Each
Quantity
Catalog number: 461120050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
71-58-9
IUPAC Name
(1R,3aS,3bR,5S,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Molecular Formula
C24H34O4
InChI Key
PSGAAPLEWMOORI-PEINSRQWSA-N
SMILES
C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(C)=O)(C(C)=O)[C@@]3(C)CC[C@@H]2[C@@]2(C)CCC(=O)C=C12
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
203°C to 209°C
Loss on drying
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text