Thermo Scientific Chemicals

Ondansetron hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 461160050
5 g, Each
Thermo Scientific Chemicals

Ondansetron hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 461160050
5 g, Each
Quantity
Catalog number: 461160050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
103639-04-9
IUPAC Name
hydrogen 9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-2,3,4,9-tetrahydro-1H-carbazol-4-one dihydrate chloride
Molecular Formula
C18H24ClN3O3
InChI Key
VRSLTNZJOUZKLX-UHFFFAOYNA-N
SMILES
[H+].O.O.[Cl-].CN1C2=C(C3=CC=CC=C13)C(=O)C(CN1C=CN=C1C)CC2
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
176°C to 180°C
Water
=<10.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text