Thermo Scientific Chemicals

Procarbazine hydrochloride

Catalog number: 461182500
250 mg, Each
Thermo Scientific Chemicals

Procarbazine hydrochloride

Catalog number: 461182500
250 mg, Each
Quantity
Catalog number: 461182500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
366-70-1
IUPAC Name
hydrogen 4-[(2-methylhydrazin-1-yl)methyl]-N-(propan-2-yl)benzamide chloride
Molecular Formula
C12H20ClN3O
InChI Key
DERJYEZSLHIUKF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CNNCC1=CC=C(C=C1)C(=O)NC(C)C
Appearance (Color)
White to tan
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %
Loss on drying
=<2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text