Pyridostigmine bromide, 98%, Thermo Scientific Chemicals
Pyridostigmine bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridostigmine bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 461190050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS101-26-8
IUPAC Name3-[(dimethylcarbamoyl)oxy]-1-methylpyridin-1-ium bromide
Molecular FormulaC9H13BrN2O2
InChI KeyVNYBTNPBYXSMOO-UHFFFAOYSA-M
SMILES[Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point153°C to 157°C
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only