Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4612600101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461260010
Price (USD)
-
Quantity:
1g
Request bulk or custom format
Chemical Identifiers
CAS78628-80-5
IUPAC Namehydrogen [(2E)-6,6-dimethylhept-2-en-4-yn-1-yl](methyl)[(naphthalen-1-yl)methyl]amine chloride
Molecular FormulaC21H26ClN
InChI KeyBWMISRWJRUSYEX-SZKNIZGXSA-N
SMILES[H+].[Cl-].CN(C\C=C\C#CC(C)(C)C)CC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Solubility(5 % in methanol) Clear solution
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link