1,8-Diazafluoren-9-one, Thermo Scientific Chemicals
1,8-Diazafluoren-9-one, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,8-Diazafluoren-9-one, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 461310010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS54078-29-4
IUPAC Name6,10-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-8-one
Molecular FormulaC11H6N2O
InChI KeyFOSUVSBKUIWVKI-UHFFFAOYSA-N
SMILESO=C1C2=C(C=CC=N2)C2=C1N=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<2 %
Dye content>=70.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only