Mannose triflate, 98%
Mannose triflate, 98%
Mannose triflate, 98%
Mannose triflate, 98%
Thermo Scientific Chemicals

Mannose triflate, 98%

CAS: 92051-23-5 | C15H19F3O12S | 480.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461340010
Price (USD)
437.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
437.00
Each
Add to cart
Chemical Identifiers
CAS92051-23-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-12° to -19° (20°C, 589 nm) (c=1, CH3CN)
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Powder
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0433198Certificate of AnalysisMar 18, 2024461341000, 461340010
A0374616Certificate of AnalysisDec 13, 2022461341000, 461340010
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection