Thermo Scientific Chemicals

Mannose triflate, 98%, Thermo Scientific Chemicals

Catalog number: 461341000
100 mg, Each
Thermo Scientific Chemicals

Mannose triflate, 98%, Thermo Scientific Chemicals

Catalog number: 461341000
100 mg, Each
Quantity
Catalog number: 461341000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92051-23-5
Infrared spectrum
Conforms
Specific optical rotation
-12° to -19° (20°C, 589 nm) (c=1, CH3CN)
Appearance (Color)
White to off-white
HPLC
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text