Thermo Scientific Chemicals

Ethosuximide, Thermo Scientific Chemicals

Catalog number: 461400050
5 g, Each
Thermo Scientific Chemicals

Ethosuximide, Thermo Scientific Chemicals

Catalog number: 461400050
5 g, Each
Quantity
Catalog number: 461400050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-67-8
IUPAC Name
3-ethyl-3-methylpyrrolidine-2,5-dione
Molecular Formula
C7H11NO2
InChI Key
HAPOVYFOVVWLRS-UHFFFAOYNA-N
SMILES
CCC1(C)CC(=O)NC1=O
Appearance (Form)
Powder or chunks
Water
=<0.5 %
Infrared spectrum
Conforms
Melting point
47°C to 52°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text