Thermo Scientific Chemicals

Chalcogran, 96%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 461410010
1 g, Each
Thermo Scientific Chemicals

Chalcogran, 96%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 461410010
1 g, Each
Quantity
Catalog number: 461410010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38401-84-2
IUPAC Name
(2S,5R)-2-ethyl-1,6-dioxaspiro[4.4]nonane
Molecular Formula
C9H16O2
InChI Key
DCWKALWZHORJAO-DTWKUNHWSA-N
SMILES
CC[C@H]1CC[C@@]2(CCCO2)O1
Appearance (Color)
Clear colorless
Refractive index
1.4420 to 1.4465 (20°C, 589 nm)
GC
>=95.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text