Chemical Identifiers
CAS637-32-1
IUPAC Namehydrogen 1-({amino[(4-chlorophenyl)amino]methylidene}amino)-N'-(propan-2-yl)methanimidamide chloride
Molecular FormulaC11H17Cl2N5
InChI KeySARMGXPVOFNNNG-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(C)N=C(N)N=C(N)NC1=CC=C(Cl)C=C1
View more