Thermo Scientific Chemicals

Atracurium besylate, 96%

Catalog number: 461470050
5 g, Each
Thermo Scientific Chemicals

Atracurium besylate, 96%

Catalog number: 461470050
5 g, Each
Quantity
Catalog number: 461470050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64228-81-5
IUPAC Name
1-[(3,4-dimethoxyphenyl)methyl]-2-[3-({5-[(3-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium-2-yl}propanoyl)oxy]pentyl}oxy)-3-oxopropyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium dibenzenesulfonate
Molecular Formula
C65H82N2O18S2
InChI Key
XXZSQOVSEBAPGS-UHFFFAOYNA-L
SMILES
[O-]S(=O)(=O)C1=CC=CC=C1.[O-]S(=O)(=O)C1=CC=CC=C1.COC1=CC=C(CC2C3=CC(OC)=C(OC)C=C3CC[N+]2(C)CCC(=O)OCCCCCOC(=O)CC[N+]2(C)CCC3=CC(OC)=C(OC)C=C3C2CC2=CC=C(OC)C(OC)=C2)C=C1OC
Appearance (Color)
White to yellowish-white
Appearance (Form)
Powder
HPLC
>=95.0 % (mix of isomers)
Infrared spectrum
Conforms
Water
=<5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text