Thermo Scientific Chemicals

Prasugrel, 96%, Thermo Scientific Chemicals

Catalog number: 461480010
1 g, Each
Thermo Scientific Chemicals

Prasugrel, 96%, Thermo Scientific Chemicals

Catalog number: 461480010
1 g, Each
Quantity
Catalog number: 461480010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
150322-43-3
IUPAC Name
5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4H,5H,6H,7H-thieno[3,2-c]pyridin-2-yl acetate
Molecular Formula
C20H20FNO3S
InChI Key
DTGLZDAWLRGWQN-UHFFFAOYNA-N
SMILES
CC(=O)OC1=CC2=C(CCN(C2)C(C(=O)C2CC2)C2=CC=CC=C2F)S1
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=95 %
Water
=<2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text