Thermo Scientific Chemicals

Hydroquinine, 95%, Thermo Scientific Chemicals

Catalog number: 461650010
1 g, Each
Thermo Scientific Chemicals

Hydroquinine, 95%, Thermo Scientific Chemicals

Catalog number: 461650010
1 g, Each
Quantity
Catalog number: 461650010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
522-66-7
IUPAC Name
(R)-{5-ethyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol
Molecular Formula
C20H26N2O2
InChI Key
LJOQGZACKSYWCH-ADMHDHIONA-N
SMILES
CCC1CN2CCC1CC2[C@H](O)C1=CC=NC2=CC=C(OC)C=C12
Appearance (Color)
White to light yellow to grey
Melting point
168°C to 176°C
HPLC
>=94 %
Appearance (Form)
Powder or chunks
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text