Caffeine, citrated
Caffeine, citrated
Caffeine, citrated
Thermo Scientific Chemicals

Caffeine, citrated

Caffeine, citrated, Quantity: 100g, Melting Point: 159.0 deg.C to 161.0 deg.C, Molecular Weight: 386.31, CAS: 69-22-7, Formula Weight: 386.31, Health Hazard 1: GHS Signal Word: Warning, Health Hazard 2: GHS H Statement Harmful if swallowed. Causes serious eye irritation. | CAS: 69-22-7 | C14H18N4O9 | 386.32 g/mol
Have Questions?
Catalog NumberQuantity
461721000100 g
Catalog number 461721000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS69-22-7
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC14H18N4O9
InChI KeyRCQXSQPPHJPGOF-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O.CN1C=NC2=C1C(=O)N(C)C(=O)N2C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<5 %
Infrared spectrumConforms
Titration with HClO448 to 52 % (Caffeine)
Titration with TBAH48 to 52 % (Citric acid)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link