Chlorothiazide, 98%, Thermo Scientific Chemicals
Chlorothiazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorothiazide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 461740050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS58-94-6
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only