Thermo Scientific Chemicals

Cilnidipine, 99%

Catalog number: 461750010
1 g, Each
Thermo Scientific Chemicals

Cilnidipine, 99%

Catalog number: 461750010
1 g, Each
Quantity
Catalog number: 461750010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
132203-70-4
IUPAC Name
3-(2-methoxyethyl) 5-(2E)-3-phenylprop-2-en-1-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C27H28N2O7
InChI Key
KJEBULYHNRNJTE-DHZHZOJONA-N
SMILES
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OCC=CC1=CC=CC=C1
Appearance (Color)
Off-white to light yellow
Infrared spectrum
Conforms
Melting point
108°C to 112°C
Loss on drying
=<1 %
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text