Thermo Scientific Chemicals

Clofazimine, 98%

Catalog number: 461760050
5 g, Each
Thermo Scientific Chemicals

Clofazimine, 98%

Catalog number: 461760050
5 g, Each
Quantity
Catalog number: 461760050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2030-63-9
IUPAC Name
N,5-bis(4-chlorophenyl)-3-[(propan-2-yl)imino]-3,5-dihydrophenazin-2-amine
Molecular Formula
C27H22Cl2N4
InChI Key
WDQPAMHFFCXSNU-UHFFFAOYSA-N
SMILES
CC(C)N=C1C=C2N(C3=CC=C(Cl)C=C3)C3=CC=CC=C3N=C2C=C1NC1=CC=C(Cl)C=C1
Appearance (Color)
Orange to red to brown
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
HPLC
>=97.5 %
Loss on drying
=<0.5 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text