Thermo Scientific Chemicals

Fenofibrate, 98%

Catalog number: 461781000
100 g, Each
Thermo Scientific Chemicals

Fenofibrate, 98%

Catalog number: 461781000
100 g, Each
Quantity
Catalog number: 461781000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
49562-28-9
IUPAC Name
propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Molecular Formula
C20H21ClO4
InChI Key
YMTINGFKWWXKFG-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
Appearance (Color)
White to off-white
Melting point
79°C to 83°C
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (60°C) (vacuum)
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text