Gemfibrozil, 98%
Gemfibrozil, 98%
Gemfibrozil, 98%
Thermo Scientific Chemicals

Gemfibrozil, 98%

CAS: 25812-30-0 | C15H22O3 | 250.34 g/mol
Have Questions?
Catalog NumberQuantity
4617900505 g
Catalog number 461790050
Price (USD)
77.60
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
77.60
Each
Add to cart
Chemical Identifiers
CAS25812-30-0
IUPAC Name5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
Molecular FormulaC15H22O3
InChI KeyHEMJJKBWTPKOJG-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point58°C to 63°C
HPLC>=97.5 %
Water=<0.25 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection