Gemfibrozil, 98%
Thermo Scientific Chemicals

Gemfibrozil, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 461790250
Price (USD)/ Each
244.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
288.00 
Save 43.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
244.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
288.00 
Save 43.35 (15%)
Add to cart
Gemfibrozil, 98%
Catalog number461790250
Price (USD)/ Each
244.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
288.00 
Save 43.35 (15%)
-
Add to cart
Chemical Identifiers
CAS25812-30-0
IUPAC Name5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
Molecular FormulaC15H22O3
InChI KeyHEMJJKBWTPKOJG-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.25 %
Appearance (Color)White to light yellow
Melting point58°C to 63°C
HPLC>=97.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only