Thermo Scientific Chemicals

Pyrantel pamoate, 98%

Catalog number: 461891000
100 g, Each
Thermo Scientific Chemicals

Pyrantel pamoate, 98%

Catalog number: 461891000
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 461891000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22204-24-6
Appearance (Form)
Powder
Titration with HClO4
97 to 103 % (on dry substance)
Appearance (Color)
Yellow to brown
Loss on drying
=<2 % (60°C, 3 h) (vacuum)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text