Triclabendazole, 98%
Triclabendazole, 98%
Thermo Scientific Chemicals

Triclabendazole, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461960010
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
Add to cart
Triclabendazole, 98%
Catalog number461960010
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.30 
Save 12.65 (15%)
-
Add to cart
Chemical Identifiers
CAS68786-66-3
IUPAC Name6-chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfanyl)-1H-1,3-benzodiazole
Molecular FormulaC14H9Cl3N2OS
InChI KeyNQPDXQQQCQDHHW-UHFFFAOYSA-N
SMILESCSC1=NC2=CC(OC3=CC=CC(Cl)=C3Cl)=C(Cl)C=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystallinje powder or crystals
Infrared spectrumConforms
Loss on drying=<1 % (1 g, 105°C, 6 h)
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only