Triclabendazole, 98%
Triclabendazole, 98%
Thermo Scientific Chemicals

Triclabendazole, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461960050
Price (USD)/ Each
285.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
285.00
Add to cart
Triclabendazole, 98%
Catalog number461960050
Price (USD)/ Each
285.00
-
Add to cart
Chemical Identifiers
IUPAC Name6-chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfanyl)-1H-1,3-benzodiazole
Molecular FormulaC14H9Cl3N2OS
InChI KeyNQPDXQQQCQDHHW-UHFFFAOYSA-N
SMILESCSC1=NC2=CC(OC3=CC=CC(Cl)=C3Cl)=C(Cl)C=C2N1
Molecular Weight (g/mol)359.65
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point175°C to 180°C
Appearance (Color)White to light yellow
Appearance (Form)Crystallinje powder or crystals
Infrared spectrumConforms
Loss on drying=<1 % (1 g, 105°C, 6 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only