Thermo Scientific Chemicals

Xylometazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 461990050
5 g, Each
Thermo Scientific Chemicals

Xylometazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 461990050
5 g, Each
Quantity
Catalog number: 461990050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1218-35-5
IUPAC Name
hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
Molecular Formula
C16H25ClN2
InChI Key
YGWFCQYETHJKNX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 % (on dry substance)
Loss on drying
=<0.5 % (105°C, 4 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text