Thermo Scientific Chemicals

1,2-Bis(trichlorosilyl)ethane, Thermo Scientific Chemicals

Catalog number: 462030250
25 g, Each
Thermo Scientific Chemicals

1,2-Bis(trichlorosilyl)ethane, Thermo Scientific Chemicals

Catalog number: 462030250
25 g, Each
Quantity
Catalog number: 462030250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2504-64-5
IUPAC Name
trichloro[2-(trichlorosilyl)ethyl]silane
Molecular Formula
C2H4Cl6Si2
InChI Key
WDVUXWDZTPZIIE-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl
Appearance (Color)
Colorless to light yellow
Appearance (Form)
Crystalline mass or liquid
Infrared spectrum
Conforms
Refractive index
1.4735 to 1.4760 (20°C, 589 nm)
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text